siRNA Details
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012225.2 | NP_001012225.1 | 16 | 2138 | ||
NM_001160155.1 | NP_001153627.1 | 16 | 2165 | |||
XM_006244840.2 | XP_006244902.1 | 16 | 2175 | |||
XM_008767040.1 | XP_008765262.1 | 15 | 1853 |

More Information
Other Species Targeted

Gene Ontology Categories:
