siRNA Details
Gene Symbol: Fam181b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109150.2 | NP_001102620.1 | 1 | 202 | ||
GenBank mRNA | BC158703.1 | - | 1 | 202 | 9029854 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam181b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109150.2 | NP_001102620.1 | 1 | 202 | ||
GenBank mRNA | BC158703.1 | - | 1 | 202 | 9029854 |