siRNA Details
Gene Symbol: Fam181b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109150.2 | NP_001102620.1 | 1 | 1244 | ||
GenBank mRNA | BC158703.1 | - | 1 | 1244 | 9029854 |

More Information
Other Species Targeted

