siRNA Details
Gene Symbol: LOC499219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047938.1 | NP_001041403.1 | 1 | 73 | ||
GenBank mRNA | AY539872.1 | - | 1 | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
