siRNA Details
Gene Symbol: RGD1562433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077678.1 | NP_001071146.1 | 1 | 647 | ||
GenBank mRNA | BC127523.1 | - | 1 | 647 | 7113805 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1562433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077678.1 | NP_001071146.1 | 1 | 647 | ||
GenBank mRNA | BC127523.1 | - | 1 | 647 | 7113805 |