siRNA Details
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759845.1 | XP_008758067.1 | 3 | 1805 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759845.1 | XP_008758067.1 | 3 | 1805 |