siRNA Details
Gene Symbol: RGD1559600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109153.1 | NP_001102623.1 | 2 | 462 | ||
XM_006230140.2 | XP_006230202.1 | 2 | 2636 | |||
XM_006230141.2 | XP_006230203.1 | 1 | 413 |
