siRNA Details
Gene Symbol: Prss53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109156.1 | NP_001102626.1 | 6 | 879 | ||
XM_006230322.2 | XP_006230384.1 | 6 | 1520 | |||
XM_006230324.2 | XP_006230386.1 | 6 | 1550 | |||
XM_008759914.1 | XP_008758136.1 | 5 | 1668 | |||
GenBank mRNA | BC161977.1 | - | 5 | 989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
