siRNA Details
Gene Symbol: Mapk1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122782.1 | NP_001116254.1 | 1 | 1254 | ||
XM_006230487.2 | XP_006230549.1 | 3 | 2398 | |||
XM_006230488.2 | XP_006230550.1 | 2 | 2689 | |||
XM_006230489.2 | XP_006230551.1 | 4 | 1563 | |||
XM_006230490.2 | XP_006230552.1 | 3 | 1440 | |||
XM_006230491.2 | XP_006230553.1 | 2 | 1319 | |||
XM_008760006.1 | XP_008758228.1 | 3 | 1854 | |||
XM_008760007.1 | XP_008758229.1 | 2 | 1731 | |||
GenBank mRNA | BC158819.1 | - | 1 | 1254 | 7374980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
