siRNA Details
Gene Symbol: Lrrc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143756.1 | NP_001137228.1 | 11 | 1629 | ||
XM_006230495.2 | XP_006230557.1 | 11 | 1703 | |||
XM_008760008.1 | XP_008758230.1 | 11 | 2531 | |||
XM_008760009.1 | XP_008758231.1 | 10 | 1551 | |||
GenBank mRNA | BC158629.1 | - | 10 | 1527 | 7456037 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
