siRNA Details
Gene Symbol: LOC499306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024291.1 | NP_001019462.1 | 5 | 468 | ||
GenBank mRNA | BC078985.1 | - | 5 | 468 | 7112339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
