siRNA Details
Gene Symbol: LOC100910973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FM045317.1 | - | 2 | 119 | ||
FM097063.1 | - | 2 | 137 | |||
FM098643.1 | - | 2 | 309 | |||
FM110991.1 | - | 2 | 112 | |||
FQ212263.1 | - | 2 | 133 | |||
FQ220597.1 | - | 2 | 257 | |||
FQ230677.1 | - | 2 | 133 | |||
FQ230710.1 | - | 2 | 133 | |||
FQ231880.1 | - | 2 | 131 | |||
FQ234947.1 | - | 2 | 129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
