siRNA Details
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037793.2 | NP_001032882.2 | 4 | 513 | ||
XM_006231244.2 | XP_006231306.1 | 4 | 724 | |||
XM_006231245.2 | XP_006231307.1 | 3 | 1288 | |||
XM_008760310.1 | XP_008758532.1 | 5 | 779 | |||
XM_008760311.1 | XP_008758533.1 | 4 | 365 |
