siRNA Details
Gene Symbol: LOC499339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025036.1 | NP_001020207.1 | 4 | 424 | ||
XM_006231246.2 | XP_006231308.1 | 4 | 471 | |||
GenBank mRNA | BC092579.1 | - | 4 | 424 | 7323921 | |
BC098744.1 | - | 3 | 398 | 7387202 |
