siRNA Details
Gene Symbol: LOC100912218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749135.3 | XP_003749183.1 | 1507 | |||
XM_006231593.2 | XP_006231655.1 | 2 | 2634 | |||
XM_006231596.2 | XP_006231658.1 | 2 | 2634 |

More Information
Other Species Targeted

