siRNA Details
Gene Symbol: Emx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109169.1 | NP_001102639.1 | 3 | 1514 | ||
XM_008760545.1 | XP_008758767.1 | 2 | 1305 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Emx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109169.1 | NP_001102639.1 | 3 | 1514 | ||
XM_008760545.1 | XP_008758767.1 | 2 | 1305 |