siRNA Details
Gene Symbol: Emx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109169.1 | NP_001102639.1 | 2, 3 | 1440 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010132, AK132093, X68882, AY117415.
Gene Ontology Categories:
