siRNA Details
Gene Symbol: Gemin7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109170.1 | NP_001102640.1 | 3 | 600 | ||
XM_006228460.2 | XP_006228522.1 | 2 | 840 | |||
XM_006228461.2 | XP_006228523.1 | 2 | 774 |
Gene Symbol: Gemin7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749742.2 | XP_003749790.1 | 1 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
