siRNA Details
Gene Symbol: Znrd1as1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166303.1 | NP_001159775.1 | 3 | 763 | ||
XM_006255877.2 | XP_006255939.1 | 3 | 1626 | |||
XM_008772699.1 | XP_008770921.1 | 3 | 844 | |||
GenBank mRNA | FQ213683.1 | - | 2 | 874 | ||
FQ214505.1 | - | 3 | 823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
