siRNA Details
Gene Symbol: Zfp365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025145.1 | NP_001020316.1 | 3 | 1017 | ||
GenBank mRNA | BC087056.1 | - | 3 | 1017 | 7124000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
