siRNA Details
Gene Symbol: Lrrc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109171.1 | NP_001102641.1 | 4 | 396 | ||
XM_006256465.2 | XP_006256527.1 | 4 | 419 | |||
XM_006256466.2 | XP_006256528.1 | 4 | 1211 | |||
XM_006256467.2 | XP_006256529.1 | 4 | 440 | |||
XM_006256468.2 | XP_006256530.1 | 4 | 452 | |||
XM_008772902.1 | XP_008771124.1 | 3 | 218 | |||
XM_008772903.1 | XP_008771125.1 | 3 | 1185 | |||
GenBank mRNA | FQ223562.1 | - | 4 | 390 |
