siRNA Details
Gene Symbol: Atp5g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133556.2 | NP_598240.1 | 5 | 585 | ||
GenBank mRNA | BC128726.1 | - | 5 | 585 | 7321526 | |
D13124.1 | - | 4 | 555 | |||
FQ209535.1 | - | 594 | ||||
FQ223576.1 | - | 5 | 598 | |||
FQ235238.1 | - | 5 | 595 |
Gene Symbol: RGD1566212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760711.1 | XP_008758933.1 | 4 | 876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
