siRNA Details
Gene Symbol: Ccdc125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134761.1 | NP_001128233.1 | 11 | 1427 | ||
GenBank mRNA | BC166734.1 | - | 7 | 1286 | 7936477 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134761.1 | NP_001128233.1 | 11 | 1427 | ||
GenBank mRNA | BC166734.1 | - | 7 | 1286 | 7936477 |