siRNA Details
Gene Symbol: LOC499541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047942.1 | NP_001041407.1 | 2 | 218 | ||
XM_008760791.1 | XP_008759013.1 | 5 | 1146 | |||
GenBank mRNA | AY383700.1 | - | 2 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
