siRNA Details
Gene Symbol: Zfp458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024299.1 | NP_001019470.1 | 4 | 2284 | ||
NM_001142962.1 | NP_001136434.1 | 4 | 2185 | |||
XM_008760843.1 | XP_008759065.1 | 5 | 2518 | |||
GenBank mRNA | BC085784.1 | - | 4 | 2284 | 7107819 |
