siRNA Details
Gene Symbol: RGD1565641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109179.1 | NP_001102649.1 | 2 | 151 | ||
XM_006232114.2 | XP_006232176.1 | 2 | 294 | |||
GenBank mRNA | FQ222537.1 | - | 2 | 128 | ||
FQ222837.1 | - | 2 | 159 |

More Information
Other Species Targeted

