siRNA Details
Gene Symbol: LOC499573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047947.1 | NP_001041412.1 | 1 | 71 | ||
GenBank mRNA | AY383711.1 | - | 1 | 71 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC499573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047947.1 | NP_001041412.1 | 1 | 71 | ||
GenBank mRNA | AY383711.1 | - | 1 | 71 |