siRNA Details
Gene Symbol: Rfxap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044263.1 | NP_001037728.1 | 643 | |||
GenBank mRNA | BC098816.1 | - | 643 | 7377196 |
Gene Symbol: Rfxapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761021.1 | XP_008759243.1 | 2 | 670 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
