siRNA Details
Gene Symbol: Tsc22d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191960.1 | NP_001178889.1 | 4 | 2141 | ||
XM_006232386.2 | XP_006232448.1 | 3 | 2970 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081229, AK007760, AK156074, BC058221, EU004151, and others.
Gene Ontology Categories:
