siRNA Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109190.2 | NP_001102660.1 | 8 | 1661 | ||
XM_006232897.2 | XP_006232959.1 | 8 | 1587 | |||
XM_006232898.2 | XP_006232960.1 | 8 | 1585 | |||
XM_006232900.2 | XP_006232962.1 | 8 | 1584 | |||
XM_006232903.2 | XP_006232965.1 | 7 | 1958 | |||
XM_006232904.2 | XP_006232966.1 | 9 | 3315 | |||
XM_006232905.2 | XP_006232967.1 | 9 | 3348 | |||
XM_006232906.2 | XP_006232968.1 | 7 | 4420 | |||
XM_006232907.2 | XP_006232969.1 | 7 | 905 | |||
XM_006232911.2 | XP_006232973.1 | 7 | 782 | |||
XM_006232912.2 | XP_006232974.1 | 6 | 704 | |||
XM_008761322.1 | XP_008759544.1 | 9 | 3902 | |||
XM_008761323.1 | XP_008759545.1 | 8 | 871 | |||
XM_008761324.1 | XP_008759546.1 | 7 | 3025 | |||
XM_008761325.1 | XP_008759547.1 | 7 | 2979 | |||
XM_008761326.1 | XP_008759548.1 | 7 | 2976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
