siRNA Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109190.2 | NP_001102660.1 | 1 | 849 | ||
XM_006232897.2 | XP_006232959.1 | 1 | 775 | |||
XM_006232898.2 | XP_006232960.1 | 1 | 776 | |||
XM_006232900.2 | XP_006232962.1 | 1 | 775 | |||
XM_006232902.2 | XP_006232964.1 | 1 | 775 | |||
XM_006232904.2 | XP_006232966.1 | 2 | 2503 | |||
XM_006232905.2 | XP_006232967.1 | 2 | 2536 | |||
XM_006232908.2 | XP_006232970.1 | 1 | 773 | |||
XM_008761322.1 | XP_008759544.1 | 2 | 3090 |

More Information
Other Species Targeted

Gene Ontology Categories:
