siRNA Details
Gene Symbol: Lysmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024302.1 | NP_001019473.1 | 2 | 686 | ||
GenBank mRNA | BC089113.1 | - | 2 | 686 | 7319992 | |
BC092612.1 | - | 2 | 48 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
