siRNA Details
Gene Symbol: Gstm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024304.1 | NP_001019475.1 | 1, 2 | 137 | ||
GenBank mRNA | BC091199.1 | - | 1, 2 | 137 | 7379741 | |
FJ179403.1 | - | 1, 2 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
