siRNA Details
Gene Symbol: RGD1560065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134616.1 | NP_001128088.1 | 2 | 306 | ||
XM_006233461.2 | XP_006233523.1 | 2 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
