siRNA Details
Gene Symbol: RGD1564379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126295.1 | NP_001119767.1 | 2 | 1582 | ||
XM_006233690.2 | XP_006233752.1 | 2 | 1654 | |||
XM_006233691.2 | XP_006233753.1 | 2 | 1582 | |||
XM_006233692.1 | XP_006233754.1 | 2 | 1408 | |||
GenBank mRNA | BC161868.1 | - | 2 | 1582 | 7108986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
