siRNA Details
Gene Symbol: LOC499770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047110.1 | NP_001040575.1 | 4, 5 | 476 | ||
XM_006233840.2 | XP_006233902.1 | 7, 8 | 1150 | |||
XM_006233841.2 | XP_006233903.1 | 7, 8 | 939 | |||
XM_006233842.2 | XP_006233904.1 | 7, 8 | 727 | |||
XM_008761672.1 | XP_008759894.1 | 8, 9 | 803 | |||
GenBank mRNA | BC098818.1 | - | 4, 5 | 476 | 7377299 |
