siRNA Details
Gene Symbol: Swi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001246661.1 | NP_001233590.1 | 5 | 517 | ||
GenBank mRNA | BC082799.1 | - | 5 | 517 | 7304460 | |
FQ221000.1 | - | 5 | 567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
