siRNA Details
Gene Symbol: Ttc30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127607.1 | NP_001121079.1 | 1 | 1201 | ||
GenBank mRNA | BC166739.1 | - | 1 | 1201 | 7937387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
