siRNA Details
Gene Symbol: LOC499843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047953.1 | NP_001041418.1 | 6, 7 | 541 | ||
GenBank mRNA | AY387077.1 | - | 6, 7 | 541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
