siRNA Details
Gene Symbol: Fibin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025042.2 | NP_001020213.1 | 1 | 673 | ||
GenBank mRNA | BC085873.1 | - | 1 | 617 | 7191397 | |
CK364593.1 | - | 1 | 673 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fibin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025042.2 | NP_001020213.1 | 1 | 673 | ||
GenBank mRNA | BC085873.1 | - | 1 | 617 | 7191397 | |
CK364593.1 | - | 1 | 673 |