siRNA Details
Gene Symbol: Fibin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025042.2 | NP_001020213.1 | 1 | 959 | ||
GenBank mRNA | BC085873.1 | - | 1 | 903 | 7191397 |

More Information
Other Species Targeted

