siRNA Details
Gene Symbol: Zfyve19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034948.1 | NP_001030120.1 | 5, 6 | 576 | ||
XM_006234786.2 | XP_006234848.1 | 5, 6 | 512 | |||
XM_006234787.2 | XP_006234849.1 | 5, 6 | 512 | |||
XM_008762106.1 | XP_008760328.1 | 5, 6 | 576 | |||
GenBank mRNA | BC099795.1 | - | 5, 6 | 576 | 7936116 |
