siRNA Details
Gene Symbol: RGD1565616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109206.1 | NP_001102676.1 | 36 | 3472 | ||
XM_008762193.1 | XP_008760415.1 | 30 | 3025 |

More Information
Other Species Targeted

