siRNA Details
Gene Symbol: Slx4ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271320.1 | NP_001258249.1 | 5, 6 | 496 | ||
XM_008762255.1 | XP_008760477.1 | 5, 6 | 370 | |||
XM_008762256.1 | XP_008760478.1 | 6, 7 | 768 | |||
XM_008762257.1 | XP_008760479.1 | 5, 6 | 630 | |||
XM_008762258.1 | XP_008760480.1 | 5, 6 | 367 | |||
XM_008762259.1 | XP_008760481.1 | 3, 4 | 571 | |||
XM_008762260.1 | XP_008760482.1 | 2, 3 | 556 | |||
XM_008762261.1 | XP_008760483.1 | 4, 5 | 542 |
