siRNA Details
Gene Symbol: Abhd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024314.1 | NP_001019485.1 | 9 | 987 | ||
GenBank mRNA | BC078918.1 | - | 9 | 987 | 7096446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
