siRNA Details
Gene Symbol: Abhd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024314.1 | NP_001019485.1 | 9 | 980 | ||
GenBank mRNA | BC078918.1 | - | 9 | 980 | 7096446 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abhd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024314.1 | NP_001019485.1 | 9 | 980 | ||
GenBank mRNA | BC078918.1 | - | 9 | 980 | 7096446 |