siRNA Details
Gene Symbol: Dsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025768.1 | NP_001020939.1 | 4 | 432 | ||
XM_006235375.2 | XP_006235437.1 | 4 | 742 | |||
XM_006235376.2 | XP_006235438.1 | 4 | 1413 | |||
XM_006235377.2 | XP_006235439.1 | 4 | 1505 | |||
XM_006235378.2 | XP_006235440.1 | 4 | 742 | |||
GenBank mRNA | BC098027.1 | - | 4 | 432 | 7456020 |
