siRNA Details
Gene Symbol: Ube2v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110345.2 | NP_001103815.2 | 3 | 238 | ||
GenBank mRNA | CO405669.1 | - | 3 | 213 |
Gene Symbol: LOC100912618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751325.3 | XP_003751373.1 | 1 | 201 |
Gene Symbol: LOC679539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751324.3 | XP_003751372.1 | 1 | 201 |

More Information
Other Species Targeted

Gene Ontology Categories:
