siRNA Details
Gene Symbol: RGD1564340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134620.1 | NP_001128092.1 | 2 | 923 | ||
XM_006235792.2 | XP_006235854.1 | 2 | 1868 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
