siRNA Details
Gene Symbol: Uckl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109212.1 | NP_001102682.1 | 12, 13 | 1345 | ||
XM_006235793.2 | XP_006235855.1 | 12, 13 | 4500 | |||
XM_006235794.2 | XP_006235856.1 | 12, 13 | 4497 | |||
XM_006235795.2 | XP_006235857.1 | 12, 13 | 4497 | |||
XM_006235796.2 | XP_006235858.1 | 12, 13 | 1962 | |||
XM_006235797.2 | XP_006235859.1 | 12, 13 | 1959 | |||
GenBank mRNA | BC105750.1 | - | 11, 12 | 1292 |

More Information
Other Species Targeted

Gene Ontology Categories:
